|
Fisherbrand® Circulators
Modern microprocessor technology and
PID-control.
- User friendly 3 button
menu
- Innovative, high power vario pump (up to 20 l
min-1
, 400 mbar) and 5 selectable power levels, tempering from room
temperature without additional cooling (open bath and power level 1),
infinitely adjustable between internal and external circulation, minimal
magnetic leakage field
- Large display, easy to read
- Automatic self testing
- Easy calibrating (ISO 9000)
- Safety through under level guard and
adjustable overheat protection, therefore suitable for flammable and
nonflammable liquids (saftey class FL)
- Audible alarm at low level and
overheating
|
|
|
|
Immersion Circulator
FBH 600
Immersion circulators for baths. Temperature range up
to 120°C.
-
Easy-to-read LED with numbers and
symbols to show operating mode
- Suitable for baths up to 50 l volume and
max. 25 mm wall thickness
- Upgradable with accessory kit with cooling
tubing and pump connection nozzle
- Heating capacity 1.5 kW
|
|
| Fisherbrand® Immersion Circulator FBH 600
|
|
Type
|
PU pcs. |
Item No.
|
EUR
|
| FBH 600 |
1 |
3543880 |
Price Info |
|