 |
| A question ? |
|
| |
| |
We will help you! |
| |
|
| Do you have questions ? |
 | Tel | : | +380 44 494 42 42 |
| Fax | : | +380 44 494 42 42 |
| info@hlr.com.ua |
| |
| ХИМЛАБОРРЕАКТИВ ООО |
 | ул. Щелковская, 8 |
| 07400 Бровары, Киевская обл. |
| Tel | : | +380 44 494 42 42 |
| Fax | : | +380 44 494 42 42 |
| |
Technical Service Life Science |
 | Tel | : | +380 44 593-77-70 |
| |
|
 |
|
 |
|
|
| |
 |
Series SG |
Fisherbrand® Portable Balances
Series SG
- From 1 mg resolution
- Low costs
- Automatic calibration with external weight
- Large, easy-to-read LCD display, illuminated
- Compact design
- Functions: conversion of units, counting function
- Percentage weighing, freezing of display, overall weight and specific weight
- Optionally RS 232 or USB interface available
(version "R" or "U")
- 2 years guarantee
|
|
| Fisherbrand® Portable Balances Series SG |
| Model |
Specifications
|
Item No. |
EUR |
| SG-123 |
120g x 1mg |
3128004 |
Price Info |
| SG-123R |
ditto |
3128005 |
| SG-123U |
ditto |
3128006 |
| SG-212 |
210g x 0.01g |
3128010 |
| SG-212R |
ditto |
3128011 |
| SG-212U |
ditto |
3128012 |
| SG-412 |
410gx 0.01g |
3128013 |
| SG-412R |
ditto |
3128014 |
| SG-412U |
ditto |
3128015 |
| SG-612 |
610g x 0.01g |
3128025 |
| SG-612R |
ditto |
3128026 |
| SG-612U |
ditto |
3128027 |
| SG-601 |
600g x 0.1g |
3128022 |
| SG-601R |
ditto |
3128023 |
| SG-601U |
ditto |
3128024 |
| SG-2001 |
2000g x 0.1g |
3128007 |
| SG-2001R |
ditto |
3128008 |
| SG-2001U |
ditto |
3128009 |
| SG-5001 |
5000g x 0.1g |
3128017 |
| SG-5001R |
ditto |
3128018 |
| SG-5001U |
ditto |
3128019 |
| SG-5 |
5000g x 1.0g |
3128016 |
| SG-5R |
ditto |
3128020 |
| SG-5U |
ditto |
3128021 |
| SG-10 |
10000g x 1.0g |
3128001 |
| SG-10R |
ditto |
3128002 |
| SG-10U |
ditto |
3128003 |
|
|
Fisherbrand® Balances
Series FDP
- Long service life in operation without mains thanks to integrated rechargeable battery
- Standard applications integrated: counting, percentage weighing, various weighing units
- Simple operation, also no problem for inexperienced users
- Large weighing dish for bulky containers
- Particularly robust construction
- RS 232 interface optional (version "R")
- Outstanding price/performance ratio
|
Series FDP
|
|
| Fisherbrand® Balances Series FDP |
| Model |
Specifications
|
Item No. |
EUR |
| FDP-300 |
300g x 0.01g |
3120979 |
Price Info |
| FDP-300R |
300g x 0.01g |
3120983 |
| FDP-600 |
600g x 0.02g |
3120980 |
| FDP-600R |
600g x 0.02g |
3120984 |
| FDP-1500 |
1500g x 0.05g |
3120981 |
| FDP-1500R |
1500g x 0.05g |
3120985 |
| FDP-3000 |
3000gx 0.1g |
3120982 |
| FDP-3000R |
3000gx 0.1g |
3120986 |
|
|
|
|
ХИМЛАБОРРЕАКТИВ ООО - ул. Щелковская, 8 - 07400 Бровары, Киевская обл. - Tel +380 44 494 42 42 - Fax. +380 44 494 42 42 - info@hlr.com.ua Copyright © 2011 Thermo Fisher Scientific Inc.
|
| |