 |
| A question ? |
|
| |
| |
We will help you! |
| |
|
| Do you have questions ? |
 | Tel | : | +380 44 494 42 42 |
| Fax | : | +380 44 494 42 42 |
| info@hlr.com.ua |
| |
| ХИМЛАБОРРЕАКТИВ ООО |
 | ул. Щелковская, 8 |
| 07400 Бровары, Киевская обл. |
| Tel | : | +380 44 494 42 42 |
| Fax | : | +380 44 494 42 42 |
| |
Technical Service Life Science |
 | Tel | : | +380 44 593-77-70 |
| |
|
 |
|
 |
|
|
| |
 |
|
Series Max
|
Fisherbrand® Microscopes
Series Max
Monocular and binocular microscopes for applications of standard laboratories in school, university and clinics. Integrated stand and achromatic objectives 4x, 10x, 40x (100x oil for binocular microscope). With Abbe NA1.25 condenser.
The optical unit is 45° inclinable and around 360° rotable. Mechanical movable carriage with slide holder.
The Digi Max is equipped with a high-quality 1.3 megapixels camera, software and connecting cables for PC operation, in order to present high-resolved real time pictures of the sample.
Scope of supply: Completely with 10x18 mm wide angle eyepiece, blue filter and dust cover.
|
|
| Fisherbrand® Microscopes Series Max |
|
Type
|
PU
pcs.
|
Item No.
|
EUR
|
| Digi Max Microscope with digital camera |
1 |
3146201 |
Price Info |
| Monocular Microscope Max with 4x, 10x, 100x objectives |
1 |
3130459 |
Price Info |
| Binocular Microscope Max mit 4x, 10x, 100x objectives |
1 |
3130460 |
| 2 eyepieces 15/12 mm |
1 |
3130462 |
| 2 eyepieces 5/15 mm |
1 |
3130461 |
| Eyepiece micrometer 0-18 mm/ 0.1 mm |
1 |
3130463 |
| Eyepiece micrometer scale / 0.01 mm |
1 |
3130464 |
| Cleaning Set |
1 |
3130467 |
|
Fisherbrand® Research Microscopes
Series Magnum
Magnum microscopes provide premium quality and excellent properties. Suitable for a wide range of applications in laboratories and life sciences. Available with binocular and trinocular optics. The ergonomic design with its streamlined housing and robust construction made from high performance alloy provide optimum handling on the job.
- Ocular: 10x/20 mm wide-angle
- Objective: 4x, 10x, 40x 100x (oil) semi-planachromatic lenses
- 5-fold revolver mit raster positioning
- Bright field Abbe condenser
- Probe carrier 208/138 mm with built-in mechanical plate and slide carriers
- Halogene illumination 6 V, 20 W
|
Series Magnum
|
| Fisherbrand® Research Microscopes Series Magnum |
Type
|
PU
pcs.
|
Item No.
|
EUR
|
| Magnum B Binocular microscope |
1 |
3148068 |
Price Info |
| Magnum T Trinocular microscope |
1 |
3148069 |
|
|
|
|
ХИМЛАБОРРЕАКТИВ ООО - ул. Щелковская, 8 - 07400 Бровары, Киевская обл. - Tel +380 44 494 42 42 - Fax. +380 44 494 42 42 - info@hlr.com.ua Copyright © 2011 Thermo Fisher Scientific Inc.
|
| |