Catalogue login
Username: 
Password:
Login
New User
Register here
Forgotten password?
English website
  PRODUKTSUCHE
A question ?  
  We will
help you!
Do you have questions ?
Tel:+380 44 494 42 42
Fax:+380 44 494 42 42
info@hlr.com.ua
ХИМЛАБОРРЕАКТИВ ООО
ул. Щелковская, 8
07400 Бровары, Киевская обл.
Tel:+380 44 494 42 42
Fax:+380 44 494 42 42
Technical Service
Life Science
Tel:+380 44 593-77-70
Spritzenvorsatzfilter_steril   
Fisherbrand® Syringe Filters, Sterile 
Ready-to-use syringe filters for quick, comfortable and reliable sterile filtration of small sample volumes up to 100 ml. Sterile.
  • MembraneØ: 25 mm
  • Filtration area: 3.9 cm2
  • Membrane type: mixed cellulose ester (MCE)
  • Dead volume: 0.1 ml (after air purge)
  • Max. pressure: 5 bar
  • Max. temperature: 45°C
  • Pyrogene: <0.5 Eu/ml
  • Housing material: PVC
  • Inlet: Luer lock female
  • Outlet: Luer slip male
Fisherbrand® Syringe Filters, sterile
Pore size
µm
PU
pcs.
Item No.
EUR
0.22 50 3700699 Price Info
0.45 50 3700700
Fisherbrand® Syringe Filters, Unsterile 
Ready-to-use syringe filters with different solvent resistant membranes for the sample preparation e.g. in the HPLC. For sample volumes up to 100 ml. Unsterile.
  • Membrane Ø: 25 mm
  • Filtration area: 3.9 cm2
  • Nylon, PTFE or PVDF membrane
  • Dead volume: 0.1 ml (after air purge)
  • Housing material: PE
   Spritzenvorsatzfilter_steril
Fisherbrand® Syringe Filters, unsterile
Membrane
Pore size
µm
PU
pcs.
Item No.
EUR
Nylon 0.45 100 FB34202 Price Info
PTFE 0.45 100 FB34203
PVDF 0.45 100 FB34201
Spritzenvorsatzfilter_15mm   
Fisherbrand® Syringe Filters 15 mm 
15 mm syringe filters in edge-extruded version for sample volumes from 1-10 ml.
RC-Membrane:
Clearing and degassing of HPLC eluents compatible with biological solutions (pH 3-12).
PA-Membrane:
Filtration of HPLC and GC solvents, elimination of particles and sterile filtration of alcaline solutions, high mechanical strength.
PTFE-Membrane:
Hydrophobic. Filtration of aggressive solvents, filtration of air and gases.
PVDF-Membrane:
Hydrophilic. Filtration biological solutions, low protein adsorption, not compatible with GC samples, strong acids, bases and keytones.
  • Filter Ø: 15 mm
  • Filtration area: 132 mm2
  • Housing material: polypropylene
  • Dead volume: <30 µl
  • Max. housing pressure: 8 bar
  • Inlet: Luer lock female
  • Outlet: Luer slip male
Fisherbrand® Syringe Filters 15 mm
Membrane
Pore size
µm
PU
pcs.
Item No.
EUR
RC 0.2 100 3152902 Price Info
RC 0.45 100 3152903
PA 0.2 100 3152904
PA 0.45 100 3152905
PTFE 0.2 100 3152906
PTFE 0.45 100 3152907
PVDF 0.2 100 3152908
PVDF 0.45 100 3152909
Glass fiber 1-2 100 3152910
Fisherbrand® Syringe Filters 25 mm 
Ready-to-use syringe filters for fast filtration of sample volumes up to 100 ml.
  • Filter Ø: 25 mm
  • Filtration area: 314 mm2
  • Housing material: polypropylene
  • Dead volume: <70 µl
  • Max. housing pressure: 5 bar
  • Inlet: Luer lock female
  • Outlet: Luer slip male
   Spritzenvorsatzfilter_25mm
Fisherbrand® Syringe Filters 25 mm
Membrane
Pore size
µm
PU
pcs.
Item No.
EUR
RC 0.2 100 3152911 Price Info
RC 0.45 100 3152912
PA 0.2 100 3152913
PA 0.45 100 3152914
PTFE 0.2 100 3152915
PTFE 0.45 100 3152916
PVDF 0.2 100 3152917
PVDF 0.45 100 3152918
Spritzenvorsatzfilter_25mm_steril   
Fisherbrand® Syringe Filters 25 mm, Sterile 
Sterile ready-to-use syringe filters for fast filtration of sample volumes up to 100 ml.
  • Filter Ø: 25 mm
  • Filtration area: 314 mm2
  • Housing material: polypropylene
  • Dead volume: <70 µl
  • Max. housing pressure: 5 bar
  • Inlet: Luer lock female
  • Outlet: Luer slip male
Fisherbrand® Syringe Filters 25 mm Sterile
Membrane
Pore size
µm
PU
pcs.
Item No.
EUR
RC 0.2 50 3152936 Price Info
RC 0.45 50 3152937
PA 0.2 50 3152938
PA 0.45 50 3152939
PTFE 0.2 50 3152940
PTFE 0.45 50 3152941
PVDF 0.2 50 3152942
PVDF 0.45 50 3152943
Glass fiber 1-2 50 3152944
Fisherbrand® Syringe Filters 25 mm
Edge-extruded version
Optional with Integrated Prefilter
The edge-extrusion provides a better fitting between membrane and filter housing and a higher maximum burst pressure. With the integration of a glass fibre membrane, the filtration time of particle loaded samples can be reduced. Premature blockage of the membrane can be prevented.
  • Filter Ø: 25 mm
  • Filtration area: 314 mm2
  • Housing material: polypropylene
  • Dead volume: <70 µl
  • Max. housing pressure: 7 bar
  • Inlet: Luer lock female
  • Outlet: Luer slip male
   Spritzenvorsatzfilter_25mm_raumspri
Fisherbrand® Syringe Filters 25 mm, Edge-extrude Version, Optional with Integrated Prefilter
Membrane
Pore size
µm
Prefilter
PU
pcs.
Item No.
EUR
RC 0.2 100 3152919 Price Info
RC 0.45 100 3152920
PA 0.2 100 3152921
PA 0.45 100 3152922
PTFE 0.2 100 3152923
PTFE 0.45 100 3152924
PVDF 0.2 100 3152925
PVDF 0.45 100 3152926
Glass fiber 1-2 100 3152927
RC 0.2 +GF 100 3152928 Price Info
RC 0.45 +GF 100 3152929
PA 0.2 +GF 100 3152930
PA 0.45 +GF 100 3152931
PTFE 0.2 +GF 100 3152932
PTFE 0.45 +GF 100 3152933
PVDF 0.2 +GF 100 3152934
PVDF 0.45 +GF 100 3152935
Spritzenvorsatzfilter_30mm   
Fisherbrand® Syringe Filters 30 mm
Edge-extruded version
Optional with Integrated Prefilter
For a higher sample throughput. Thanks to the bigger diameter this filter has more than 40% larger filtration area than the 25 mm filter.
Durch die Integration einer Glasfasermembran wird die Filtrationszeit von stark partikelbelasteten Medien nochmals reduziert. Ein vorzeitiges Verblocken der Membrande wird verhindert.
  • Filter Ø: 30 mm
  • Filtration area: 490 mm2
  • Housing material: polypropylene
  • Dead volume: <170 µl
  • Max. housing pressure: 8 bar
  • Inlet: Luer lock female
  • Outlet: Luer slip male
Fisherbrand® Syringe Filter 30 mm, Edge-extruded Version, Optional with Integrated Prefilter
Membrane
Pore size
µm
Prefilter
PU
pcs.
Item No.
EUR
RC 0.2 100 3152945 Price Info
RC 0.45 100 3152946
PA 0.2 100 3152947
PA 0.45 100 3152948
PTFE 0.2 100 3152948
PTFE 0.45 100 3152950
PVDF 0.2 100 3152951
PVDF 0.45 100 3152952
Glass fiber 1-2 100 3152953
RC 0.2 +GF 100 3152954 Price Info
RC 0.45 +GF 100 3152955
PA 0.2 +GF 100 3152956
PA 0.45 +GF 100 3152957
PTFE 0.2 +GF 100 3152958
PTFE 0.45 +GF 100 3152959
PVDF 0.2 +GF 100 3152960
PVDF 0.45 +GF 100 3152961
ХИМЛАБОРРЕАКТИВ ООО - ул. Щелковская, 8 - 07400 Бровары, Киевская обл. - Tel +380 44 494 42 42 - Fax. +380 44 494 42 42 - info@hlr.com.ua
Copyright © 2011 Thermo Fisher Scientific Inc.